Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPECC1L
Entrez Gene ID
23384
Gene Name
sperm antigen with calponin homology and coiled-coil domains 1 like
Gene Aliases
CYTSA, GBBB2, OBLFC1
Chromosome Location
Chr.22: 24270817 - 24417740 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145468.3 NP_001138940.3 13 - 14 3284 71
NM_001254733.1 NP_001241662.1 6 - 7 823 71
NM_015330.4 NP_056145.4 14 - 15 3388 71
GenBank mRNA AB002374.1 - 10 - 11 2612 71
AK025531.1 - 10 - 11 1692 71
AL832425.1 - 14 - 15 3307 71
AY884293.1 - 13 - 14 3212 71
BC136471.1 - 13 - 14 3154 71

Gene Symbol
SPECC1L-ADORA2A
Entrez Gene ID
101730217
Gene Name
SPECC1L-ADORA2A readthrough (NMD candidate)
Gene Aliases
-
Chromosome Location
Chr.22: 24270817 - 24442360 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_103546.1 15 - 16 3510 71
GenBank mRNA AB209022.1 - 15 - 16 3429 71

View Full Product Details