The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015323.4 | NP_056138.1 | 15 - 16 | 1861 | 108 |
GenBank mRNA | AB018319.1 | - | 15 - 16 | 1830 | 108 |
AK295934.1 | - | 15 - 16 | 1815 | 108 | |
BC028608.1 | - | 15 - 16 | 1853 | 108 | |
BC036379.2 | - | 15 - 16 | 1846 | 108 |