The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015323.4 | NP_056138.1 | 3 - 4 | 301 | 118 |
GenBank mRNA | AB018319.1 | - | 3 - 4 | 270 | 118 |
AK295934.1 | - | 3 - 4 | 255 | 118 | |
AK307651.1 | - | 3 - 4 | 286 | 118 | |
AK316453.1 | - | 3 - 4 | 553 | 118 | |
BC015377.1 | - | 3 - 4 | 288 | 118 | |
BC028608.1 | - | 3 - 4 | 280 | 118 | |
BC036379.2 | - | 3 - 4 | 286 | 118 |