The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015323.4 | NP_056138.1 | 7 - 8 | 706 | 96 |
GenBank mRNA | AB018319.1 | - | 7 - 8 | 675 | 96 |
AK295934.1 | - | 7 - 8 | 660 | 96 | |
AK307651.1 | - | 7 - 8 | 691 | 96 | |
AK316453.1 | - | 7 - 8 | 958 | 96 | |
BC015377.1 | - | 7 - 8 | 693 | 96 | |
BC028608.1 | - | 7 - 8 | 685 | 96 | |
BC036379.2 | - | 7 - 8 | 691 | 96 |