The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015323.4 | NP_056138.1 | 8 - 9 | 846 | 85 |
GenBank mRNA | AB018319.1 | - | 8 - 9 | 815 | 85 |
AK295934.1 | - | 8 - 9 | 800 | 85 | |
AK307651.1 | - | 8 - 9 | 831 | 85 | |
AK316453.1 | - | 8 - 9 | 1098 | 85 | |
BC015377.1 | - | 8 - 9 | 833 | 85 | |
BC028608.1 | - | 8 - 9 | 825 | 85 | |
BC036379.2 | - | 8 - 9 | 831 | 85 |