The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130144.2 | NP_001123616.1 | 11 - 12 | 1985 | 63 |
NM_001164266.1 | NP_001157738.1 | 11 - 12 | 1981 | 63 | |
NM_021070.4 | NP_066548.2 | 11 - 12 | 1985 | 63 | |
XM_011545032.2 | XP_011543334.1 | 11 - 12 | 2167 | 63 | |
XM_011545033.2 | XP_011543335.1 | 11 - 12 | 2166 | 63 | |
XM_017017737.1 | XP_016873226.1 | 11 - 12 | 2172 | 63 | |
GenBank mRNA | AF135960.3 | - | 11 - 12 | 1716 | 63 |
AF318354.1 | - | 9 - 10 | 1017 | 63 | |
AK024477.1 | - | 11 - 12 | 1982 | 63 | |
AK298858.1 | - | 6 - 7 | 667 | 63 | |
AK299351.1 | - | 12 - 13 | 1647 | 63 | |
BC008761.2 | - | 2 - 3 | 178 | 63 | |
BC136277.1 | - | 11 - 12 | 1791 | 63 |