The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320383.1 | NP_001307312.1 | 2 - 3 | 284 | 84 |
NM_001320384.1 | NP_001307313.1 | 2 - 3 | 284 | 84 | |
NM_004649.7 | NP_004640.4 | 2 - 3 | 284 | 84 | |
NM_198155.4 | NP_937798.4 | 2 - 3 | 284 | 84 | |
GenBank mRNA | BC002370.2 | - | 2 - 3 | 269 | 84 |
BC003587.1 | - | 2 - 3 | 215 | 84 | |
BI196781.1 | - | 2 - 3 | 159 | 84 | |
BP283346.1 | - | 2 - 3 | 284 | 84 | |
BQ647697.1 | - | 2 - 3 | 117 | 84 | |
CX788692.1 | - | 2 - 3 | 276 | 84 | |
D86061.1 | - | 2 - 3 | 215 | 84 | |
D86062.1 | - | 2 - 3 | 216 | 84 | |
U53003.1 | - | 2 - 3 | 282 | 84 | |
Y07572.1 | - | 2 - 3 | 265 | 84 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006723901.3 | XP_006723964.1 | 2 - 3 | 307 | 84 |
XM_006723902.3 | XP_006723965.1 | 2 - 3 | 296 | 84 | |
XM_006723903.3 | XP_006723966.1 | 2 - 3 | 294 | 84 | |
XM_006723904.3 | XP_006723967.1 | 2 - 3 | 295 | 84 | |
XM_017028514.1 | XP_016884003.1 | 2 - 3 | 297 | 84 |