The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_103511.1 | 1 - 2 | 461 | 132 | |
XM_017007312.1 | XP_016862801.1 | 1 - 2 | 2099 | 132 | |
XM_017007313.1 | XP_016862802.1 | 1 - 2 | 2100 | 132 | |
GenBank mRNA | BC024239.2 | - | 1 - 2 | 231 | 132 |
BG722744.1 | - | 1 - 2 | 236 | 132 | |
DC323603.1 | - | 1 - 2 | 461 | 132 |