The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242881.1 | NP_001229810.1 | 8 - 9 | 865 | 124 |
NM_001242882.1 | NP_001229811.1 | 8 - 9 | 862 | 124 | |
NM_001242883.1 | NP_001229812.1 | 5 - 6 | 576 | 124 | |
NM_018210.3 | NP_060680.2 | 8 - 9 | 865 | 124 | |
NR_040103.1 | 8 - 9 | 857 | 124 | ||
NR_040104.1 | 7 - 8 | 773 | 124 | ||
GenBank mRNA | AK001631.1 | - | 8 - 9 | 788 | 124 |
AK057767.1 | - | 6 - 7 | 736 | 124 | |
AK093771.1 | - | 8 - 9 | 783 | 124 | |
AK225129.1 | - | 8 - 9 | 787 | 124 | |
AK296758.1 | - | 8 - 9 | 746 | 124 | |
AK298917.1 | - | 4 - 5 | 393 | 124 | |
AK302117.1 | - | 5 - 6 | 461 | 124 | |
BC041028.1 | - | 8 - 9 | 796 | 124 |