Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IKZF5
Entrez Gene ID
64376
Gene Name
IKAROS family zinc finger 5
Gene Aliases
PEGASUS, ZNFN1A5
Chromosome Location
Chr.10: 122990806 - 123008850 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_073475.1 1 - 2 151 82
NR_073476.1 1 - 2 151 82
NR_073477.1 1 - 2 151 82
NR_073479.1 1 - 2 151 82
XM_006717947.3 XP_006718010.1 1 - 2 126 82
XM_017016553.1 XP_016872042.1 1 - 2 125 82
XM_017016554.1 XP_016872043.1 1 - 2 125 82
XM_017016555.1 XP_016872044.1 1 - 2 125 82
GenBank mRNA AK026626.1 - 1 - 2 96 82
AK226143.1 - 1 - 2 92 82
BC022564.1 - 1 - 2 106 82
BX647958.1 - 1 - 2 100 82
DB080324.1 - 1 - 2 151 82
DB224006.1 - 1 - 2 96 82

View Full Product Details