The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006623.3 | NP_006614.2 | 5 - 6 | 733 | 97 |
XM_011541226.1 | XP_011539528.1 | 7 - 8 | 1014 | 97 | |
XM_011541227.2 | XP_011539529.1 | 7 - 8 | 719 | 97 | |
XM_011541228.1 | XP_011539530.1 | 7 - 8 | 632 | 97 | |
XM_011541231.2 | XP_011539533.1 | 7 - 8 | 836 | 97 | |
GenBank mRNA | AF171236.1 | - | 2 - 3 | 123 | 97 |
AF171237.1 | - | 5 - 6 | 579 | 97 | |
AK093306.1 | - | 4 - 5 | 1911 | 97 | |
AK309662.1 | - | 7 - 8 | 927 | 97 | |
AK310132.1 | - | 6 - 7 | 684 | 97 | |
AK315360.1 | - | 5 - 6 | 654 | 97 | |
BC000303.2 | - | 5 - 6 | 573 | 97 | |
BC001349.2 | - | 5 - 6 | 568 | 97 | |
BC011262.1 | - | 5 - 6 | 642 | 97 | |
BC023235.1 | - | 5 - 6 | 546 | 97 | |
CR456795.1 | - | 5 - 6 | 506 | 97 | |
FJ224347.1 | - | - | 1198 | 97 |