The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195728.2 | NP_001182657.1 | 2 - 3 | 250 | 67 |
GenBank mRNA | U01824.1 | - | 2 - 3 | 171 | 67 |