Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP4
Entrez Gene ID
4134
Gene Name
microtubule associated protein 4
Gene Aliases
-
Chromosome Location
Chr.3: 47850690 - 48089279 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005265158.2 XP_005265215.1 6 - 7 2680 87
XM_011533718.1 XP_011532020.1 15 - 16 6351 87
XM_017006403.1 XP_016861892.1 14 - 15 2967 87
XM_017006404.1 XP_016861893.1 14 - 15 2967 87
XM_017006413.1 XP_016861902.1 13 - 14 3380 87
XM_017006414.1 XP_016861903.1 13 - 14 3380 87
XM_017006416.1 XP_016861905.1 11 - 12 2757 87
XM_017006421.1 XP_016861910.1 6 - 7 2680 87
GenBank mRNA CR749544.1 - 6 - 7 1092 87

View Full Product Details