The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052817.2 | NP_438112.2 | 7 - 8 | 1914 | 100 |
XM_017029239.1 | XP_016884728.1 | 7 - 8 | 6177 | 100 | |
GenBank mRNA | AF196481.1 | - | 7 - 8 | 1354 | 100 |
AY625004.1 | - | 6 - 7 | 1281 | 100 | |
BC017707.1 | - | 6 - 7 | 1308 | 100 | |
BT006663.1 | - | 6 - 7 | 1281 | 100 |