Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATPAF1
Entrez Gene ID
64756
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 1
Gene Aliases
ATP11, ATP11p
Chromosome Location
Chr.1: 46635039 - 46668427 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042546.2 NP_001036011.2 6 - 7 688 60
GenBank mRNA AK311271.1 - 2 - 3 96 60
BX438342.2 - 6 - 7 638 60

View Full Product Details