The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284389.1 | NP_001271318.1 | 2 - 3 | 410 | 72 |
XM_005270273.1 | XP_005270330.1 | 2 - 3 | 384 | 72 | |
GenBank mRNA | AK225101.1 | - | 2 - 3 | 197 | 72 |
AY820769.1 | - | 2 - 3 | 175 | 72 | |
BC001883.1 | - | 2 - 3 | 182 | 72 |