The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001656.3 | NP_001647.1 | 6 - 7 | 1128 | 64 |
NM_033227.2 | NP_150230.1 | 6 - 7 | 1128 | 64 | |
NM_033228.2 | NP_150231.1 | 6 - 7 | 1128 | 64 | |
XM_005248497.3 | XP_005248554.1 | 6 - 7 | 1090 | 64 | |
XM_017009444.1 | XP_016864933.1 | 6 - 7 | 1112 | 64 | |
GenBank mRNA | AF230397.1 | - | 6 - 7 | 1064 | 64 |
AF230398.1 | - | 6 - 7 | 1064 | 64 | |
AF230399.1 | - | 6 - 7 | 1064 | 64 | |
BC022510.1 | - | 6 - 7 | 1069 | 64 | |
L04510.1 | - | 6 - 7 | 1064 | 64 |