The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165960.1 | NP_001159432.1 | 5 - 6 | 1090 | 65 |
NM_021628.2 | NP_067641.2 | 5 - 6 | 926 | 65 | |
XM_017024921.1 | XP_016880410.1 | 4 - 5 | 869 | 65 | |
XM_017024922.1 | XP_016880411.1 | 4 - 5 | 869 | 65 | |
XM_017024923.1 | XP_016880412.1 | 4 - 5 | 869 | 65 | |
XM_017024924.1 | XP_016880413.1 | 4 - 5 | 869 | 65 | |
XM_017024925.1 | XP_016880414.1 | 4 - 5 | 869 | 65 | |
GenBank mRNA | AJ269499.1 | - | 4 - 5 | 1055 | 65 |
AK296416.1 | - | 5 - 6 | 991 | 65 | |
AK299742.1 | - | 5 - 6 | 1033 | 65 | |
AK313677.1 | - | 5 - 6 | 890 | 65 | |
BC101938.1 | - | 4 - 5 | 778 | 65 | |
BC101939.1 | - | 4 - 5 | 778 | 65 | |
BC103508.1 | - | 4 - 5 | 778 | 65 | |
BC104724.1 | - | 4 - 5 | 778 | 65 |