Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C19orf12
Entrez Gene ID
83636
Gene Name
chromosome 19 open reading frame 12
Gene Aliases
NBIA3, NBIA4, SPG43
Chromosome Location
Chr.19: 29698886 - 29715789 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256046.1 NP_001242975.1 1 - 2 422 63
NM_001282931.1 NP_001269860.1 1 - 2 422 63
NM_031448.4 NP_113636.2 1 - 2 422 63
GenBank mRNA AK057185.1 - 1 - 2 119 63
BC063518.1 - 1 - 2 101 63
BG720088.1 - 1 - 2 151 63
BG722048.1 - 1 - 2 390 63

View Full Product Details