Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C19orf12
Entrez Gene ID
83636
Gene Name
chromosome 19 open reading frame 12
Gene Aliases
NBIA3, NBIA4, SPG43
Chromosome Location
Chr.19: 29698886 - 29715789 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031726.3 NP_001026896.2 2 - 3 323 93
NM_001256046.1 NP_001242975.1 2 - 3 594 93
NM_001256047.1 NP_001242976.1 2 - 3 400 93
NM_031448.4 NP_113636.2 2 - 3 594 93
GenBank mRNA AK057185.1 - 2 - 3 291 93
AL162066.1 - 1 - 2 98 93
BC063518.1 - 2 - 3 273 93
BG722048.1 - 2 - 3 562 93
DA189248.1 - 2 - 3 400 93
DA708831.1 - 2 - 3 316 93

View Full Product Details