The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005216.4 | NP_005207.2 | 7 - 8 | 993 | 103 |
GenBank mRNA | AB062391.1 | - | 7 - 8 | 826 | 103 |
AK296041.1 | - | 6 - 7 | 759 | 103 | |
AK297009.1 | - | 7 - 8 | 866 | 103 | |
AK315633.1 | - | 7 - 8 | 973 | 103 | |
BC002594.2 | - | 7 - 8 | 900 | 103 | |
D29643.1 | - | 7 - 8 | 956 | 103 |