The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113491.1 | NP_001106963.1 | 6 - 7 | 1253 | 109 |
NM_001113492.1 | NP_001106964.1 | 6 - 7 | 1428 | 109 | |
NM_001113493.1 | NP_001106965.1 | 5 - 6 | 1414 | 109 | |
NM_001113494.1 | NP_001106966.1 | 5 - 6 | 1367 | 109 | |
NM_001113495.1 | NP_001106967.1 | 4 - 5 | 1067 | 109 | |
NM_001113496.1 | NP_001106968.1 | 4 - 5 | 662 | 109 | |
NM_001293695.1 | NP_001280624.1 | 5 - 6 | 1196 | 109 | |
NM_001293696.1 | NP_001280625.1 | 4 - 5 | 478 | 109 | |
NM_001293697.1 | NP_001280626.1 | 4 - 5 | 691 | 109 | |
NM_001293698.1 | NP_001280627.1 | 4 - 5 | 777 | 109 | |
NM_006640.4 | NP_006631.2 | 5 - 6 | 1885 | 109 | |
XM_005256962.1 | XP_005257019.1 | 4 - 5 | 535 | 109 | |
XM_006721643.2 | XP_006721706.1 | 7 - 8 | 1356 | 109 | |
XM_006721644.1 | XP_006721707.1 | 4 - 5 | 578 | 109 | |
XM_011524204.1 | XP_011522506.1 | 7 - 8 | 1453 | 109 | |
XM_011524206.1 | XP_011522508.1 | 5 - 6 | 1173 | 109 | |
XM_011524207.1 | XP_011522509.1 | 5 - 6 | 1186 | 109 | |
XM_011524208.2 | XP_011522510.1 | 4 - 5 | 444 | 109 | |
XM_017024031.1 | XP_016879520.1 | 5 - 6 | 785 | 109 | |
XM_017024032.1 | XP_016879521.1 | 4 - 5 | 784 | 109 | |
GenBank mRNA | AB023208.1 | - | 5 - 6 | 1367 | 109 |
AF123052.1 | - | 5 - 6 | 1848 | 109 | |
AF142408.1 | - | 5 - 6 | 1330 | 109 | |
AF189712.2 | - | 7 - 8 | 1380 | 109 | |
AF189713.3 | - | 6 - 7 | 1223 | 109 | |
AJ312319.1 | - | 5 - 6 | 1414 | 109 | |
AJ312320.1 | - | 4 - 5 | 456 | 109 | |
AJ312321.1 | - | 5 - 6 | 1885 | 109 | |
AJ312322.1 | - | 5 - 6 | 1356 | 109 | |
AK022493.1 | - | 4 - 5 | 444 | 109 | |
AK056495.1 | - | 4 - 5 | 1067 | 109 | |
AK290368.1 | - | 6 - 7 | 1428 | 109 | |
AK299828.1 | - | 4 - 5 | 691 | 109 | |
AK300270.1 | - | 5 - 6 | 1155 | 109 | |
AK303449.1 | - | 4 - 5 | 471 | 109 | |
AK304143.1 | - | 5 - 6 | 1268 | 109 | |
AK316473.1 | - | 4 - 5 | 777 | 109 | |
AL080131.1 | - | 1 - 2 | 72 | 109 | |
BC021192.2 | - | 5 - 6 | 1108 | 109 | |
BC054004.1 | - | 7 - 8 | 1083 | 109 | |
BT007215.1 | - | 5 - 6 | 1073 | 109 | |
CN393537.1 | - | 4 - 5 | 470 | 109 | |
DC410238.1 | - | 4 - 5 | 479 | 109 | |
KU178538.1 | - | 4 - 5 | 428 | 109 | |
KU178539.1 | - | 5 - 6 | 1073 | 109 |