The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021137.4 | NP_066960.1 | 4 - 5 | 912 | 82 |
XM_017024993.1 | XP_016880482.1 | 4 - 5 | 868 | 82 | |
GenBank mRNA | AK300584.1 | - | 3 - 4 | 354 | 82 |
AK312387.1 | - | 4 - 5 | 673 | 82 | |
BC001643.2 | - | 4 - 5 | 659 | 82 | |
BC001949.1 | - | 4 - 5 | 659 | 82 | |
BC003694.2 | - | 4 - 5 | 674 | 82 | |
BT020121.1 | - | 3 - 4 | 463 | 82 | |
CR456819.1 | - | 3 - 4 | 463 | 82 | |
M80783.1 | - | 4 - 5 | 616 | 82 |