The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019072.2 | NP_061945.1 | 7 - 8 | 836 | 117 |
XM_005248548.3 | XP_005248605.1 | 7 - 8 | 703 | 117 | |
XM_017009597.1 | XP_016865086.1 | 8 - 9 | 972 | 117 | |
GenBank mRNA | AF368281.1 | - | 7 - 8 | 846 | 117 |
AK096321.1 | - | 7 - 8 | 791 | 117 | |
BC012044.1 | - | 7 - 8 | 813 | 117 |