The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098398.1 | NP_001091868.1 | 27 - 28 | 3243 | 62 |
NM_004371.3 | NP_004362.2 | 27 - 28 | 3216 | 62 | |
GenBank mRNA | AK129830.1 | - | 1 - 2 | 61 | 62 |
BC038447.1 | - | 27 - 28 | 2938 | 62 | |
U24105.1 | - | 28 - 29 | 3288 | 62 |