The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098398.1 | NP_001091868.1 | 8 - 9 | 1107 | 73 |
NM_004371.3 | NP_004362.2 | 8 - 9 | 1107 | 73 | |
GenBank mRNA | AK307953.1 | - | 7 - 8 | 742 | 73 |
BC038447.1 | - | 8 - 9 | 802 | 73 | |
U24105.1 | - | 9 - 10 | 1179 | 73 |