Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GON4L
Entrez Gene ID
54856
Gene Name
gon-4 like
Gene Aliases
GON-4, GON4, YARP
Chromosome Location
Chr.1: 155749658 - 155859400 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282856.1 NP_001269785.1 21 - 22 4598 68
NM_001282858.1 NP_001269787.1 21 - 22 4650 68
NM_001282860.1 NP_001269789.1 21 - 22 4650 68
XM_005245284.3 XP_005245341.1 21 - 22 4590 68
XM_005245286.3 XP_005245343.1 6 - 7 2401 68
XM_006711393.3 XP_006711456.1 21 - 22 4609 68
XM_006711394.3 XP_006711457.1 21 - 22 4595 68
XM_011509658.2 XP_011507960.1 20 - 21 4490 68
XM_011509659.2 XP_011507961.1 21 - 22 4614 68
GenBank mRNA AB046826.1 - 1 - 2 1041 68
AB195687.1 - 21 - 22 4650 68
AB195688.1 - 22 - 23 3332 68
AY335490.2 - 21 - 22 4553 68
AY335491.2 - 21 - 22 4598 68
BC117557.1 - 6 - 7 2413 68

View Full Product Details