The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014913.3 | NP_055728.1 | 3 - 4 | 650 | 120 |
XM_005266656.4 | XP_005266713.1 | 3 - 4 | 504 | 120 | |
XM_011525883.2 | XP_011524185.1 | 3 - 4 | 364 | 120 | |
GenBank mRNA | AB020670.1 | - | 3 - 4 | 380 | 120 |
AK292566.1 | - | 3 - 4 | 419 | 120 | |
BC071589.1 | - | 3 - 4 | 650 | 120 |