Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSBN1
Entrez Gene ID
54665
Gene Name
round spermatid basic protein 1
Gene Aliases
ROSBIN
Chromosome Location
Chr.1: 113761832 - 113812476 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018364.4 NP_060834.2 2 - 3 1433 94
GenBank mRNA AK002082.1 - 2 - 3 1248 94
AK022166.1 - 1 - 2 612 94
AK292552.1 - 2 - 3 1433 94
CR627402.1 - 3 - 4 1026 94

View Full Product Details