The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004656.3 | NP_004647.1 | 8 - 9 | 882 | 79 |
XM_011534149.2 | XP_011532451.1 | 8 - 9 | 878 | 79 | |
XM_011534150.2 | XP_011532452.1 | 8 - 9 | 878 | 79 | |
XM_011534152.1 | XP_011532454.1 | 8 - 9 | 878 | 79 | |
GenBank mRNA | AF045581.1 | - | 8 - 9 | 694 | 79 |
AK292608.1 | - | 8 - 9 | 816 | 79 | |
AY130008.1 | - | 2 - 3 | 167 | 79 | |
BC001596.1 | - | 8 - 9 | 770 | 79 | |
D87462.2 | - | 8 - 9 | 779 | 79 | |
D88812.1 | - | 8 - 9 | 655 | 79 |