The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000425.4 | NP_000416.1 | 15 - 16 | 2145 | 70 |
NM_001143963.2 | NP_001137435.1 | 14 - 15 | 2029 | 70 | |
NM_001278116.1 | NP_001265045.1 | 16 - 17 | 2154 | 70 | |
NM_024003.3 | NP_076493.1 | 15 - 16 | 2044 | 70 | |
GenBank mRNA | AB102653.1 | - | 15 - 16 | 1930 | 70 |
AK289754.1 | - | 15 - 16 | 1961 | 70 | |
BC126229.1 | - | 15 - 16 | 2046 | 70 | |
BC136447.1 | - | 15 - 16 | 2046 | 70 | |
EF506611.1 | - | 14 - 15 | 1921 | 70 | |
M74387.1 | - | 15 - 16 | 1947 | 70 | |
M77640.1 | - | 15 - 16 | 1956 | 70 | |
X58776.1 | - | 7 - 8 | 880 | 70 | |
X59847.1 | - | 15 - 16 | 1936 | 70 |