The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143963.2 | NP_001137435.1 | 25 - 26 | 3623 | 63 |
NM_024003.3 | NP_076493.1 | 26 - 27 | 3638 | 63 | |
GenBank mRNA | EF506611.1 | - | 25 - 26 | 3515 | 63 |
M74387.1 | - | 26 - 27 | 3541 | 63 |