Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOL9
Entrez Gene ID
79707
Gene Name
nucleolar protein 9
Gene Aliases
Grc3, NET6
Chromosome Location
Chr.1: 6521347 - 6554598 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024654.4 NP_078930.3 1 - 2 491 64
XM_017002337.1 XP_016857826.1 1 - 2 429 64
XM_017002338.1 XP_016857827.1 1 - 2 234 64
XM_017002339.1 XP_016857828.1 1 - 2 48 64
GenBank mRNA AK091284.1 - 1 - 2 428 64
AK289653.1 - 1 - 2 423 64
BC105095.1 - 1 - 2 491 64
BC110849.1 - 1 - 2 400 64
BC112278.1 - 1 - 2 491 64
BC143646.1 - 1 - 2 491 64
BE383415.1 - 1 - 2 392 64
BI828678.1 - 1 - 2 393 64

View Full Product Details