Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MAP3K2
Entrez Gene ID
10746
Gene Name
mitogen-activated protein kinase kinase kinase 2
Gene Aliases
MEKK2, MEKK2B
Chromosome Location
Chr.2: 127298669 - 127343229 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006609.4 NP_006600.3 12 - 13 1289 103
GenBank mRNA AB208963.1 - 11 - 12 1195 103
AF239798.1 - 11 - 12 1220 103
AK075004.1 - 6 - 7 653 103
AK315007.1 - 13 - 14 1474 103
BC136293.1 - 12 - 13 1258 103

View Full Product Details