The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006609.4 | NP_006600.3 | 3 - 4 | 266 | 156 |
GenBank mRNA | AB208963.1 | - | 2 - 3 | 172 | 156 |
AF111105.1 | - | 3 - 4 | 267 | 156 | |
AF239798.1 | - | 2 - 3 | 197 | 156 | |
AK315007.1 | - | 4 - 5 | 450 | 156 | |
BC136293.1 | - | 3 - 4 | 235 | 156 |