The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006609.4 | NP_006600.3 | 4 - 5 | 368 | 88 |
GenBank mRNA | AB208963.1 | - | 3 - 4 | 274 | 88 |
AK315007.1 | - | 5 - 6 | 552 | 88 | |
BC136293.1 | - | 4 - 5 | 337 | 88 |