The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286556.1 | NP_001273485.1 | 2 - 3 | 772 | 74 |
NM_001320745.1 | NP_001307674.1 | 2 - 3 | 697 | 74 | |
NM_058187.4 | NP_478067.2 | 2 - 3 | 772 | 74 | |
NR_135467.1 | 2 - 3 | 828 | 74 | ||
XM_005261019.4 | XP_005261076.1 | 2 - 3 | 893 | 74 | |
XM_006724038.3 | XP_006724101.1 | 2 - 3 | 893 | 74 | |
XM_011529669.2 | XP_011527971.1 | 2 - 3 | 1024 | 74 | |
XM_017028419.1 | XP_016883908.1 | 2 - 3 | 893 | 74 | |
XM_017028420.1 | XP_016883909.1 | 3 - 4 | 813 | 74 | |
XM_017028421.1 | XP_016883910.1 | 2 - 3 | 697 | 74 | |
GenBank mRNA | AF358258.1 | - | 2 - 3 | 355 | 74 |
AK125013.1 | - | 1 - 2 | 1322 | 74 | |
AY358787.1 | - | 2 - 3 | 765 | 74 | |
BC038710.1 | - | 2 - 3 | 507 | 74 | |
BM836911.1 | - | 1 - 2 | 100 | 74 |