The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152701.4 | NP_689914.3 | 29 - 30 | 10231 | 85 |
XM_011515130.2 | XP_011513432.1 | 29 - 30 | 10263 | 85 | |
XM_011515131.2 | XP_011513433.1 | 29 - 30 | 10263 | 85 | |
XM_011515132.2 | XP_011513434.1 | 29 - 30 | 10263 | 85 | |
XM_011515133.2 | XP_011513435.1 | 29 - 30 | 10263 | 85 | |
XM_011515134.2 | XP_011513436.1 | 29 - 30 | 10263 | 85 | |
XM_011515136.2 | XP_011513438.2 | 24 - 25 | 9628 | 85 | |
XM_011515137.2 | XP_011513439.1 | 29 - 30 | 10263 | 85 | |
XM_011515138.2 | XP_011513440.1 | 29 - 30 | 10263 | 85 | |
XM_011515139.2 | XP_011513441.1 | 29 - 30 | 10263 | 85 | |
XM_011515141.2 | XP_011513443.1 | 29 - 30 | 10264 | 85 | |
XM_011515142.2 | XP_011513444.1 | 29 - 30 | 10264 | 85 | |
XM_011515143.2 | XP_011513445.1 | 29 - 30 | 10263 | 85 | |
XM_011515144.2 | XP_011513446.1 | 29 - 30 | 10263 | 85 | |
XM_011515145.2 | XP_011513447.1 | 29 - 30 | 10264 | 85 | |
XM_011515146.2 | XP_011513448.1 | 29 - 30 | 10264 | 85 | |
XM_017011767.1 | XP_016867256.1 | 29 - 30 | 10263 | 85 | |
XM_017011768.1 | XP_016867257.1 | 29 - 30 | 10263 | 85 | |
GenBank mRNA | AF501281.1 | - | 11 - 12 | 1411 | 85 |
AK128570.1 | - | 5 - 6 | 677 | 85 | |
AY204751.1 | - | 29 - 30 | 10232 | 85 | |
JF913488.1 | - | 12 - 13 | 2126 | 85 | |
JF913491.1 | - | 12 - 13 | 2126 | 85 | |
JF913492.1 | - | 12 - 13 | 2126 | 85 |