Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GBA2
Entrez Gene ID
57704
Gene Name
glucosylceramidase beta 2
Gene Aliases
AD035, NLGase, SPG46
Chromosome Location
Chr.9: 35736862 - 35749255 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011517973.2 XP_011516275.1 16 - 17 2635 69
XM_017014938.1 XP_016870427.1 16 - 17 3075 69
XM_017014939.1 XP_016870428.1 16 - 17 2569 69
XM_017014941.1 XP_016870430.1 15 - 16 2838 69
GenBank mRNA AB046825.1 - 16 - 17 3020 69

View Full Product Details