Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDC16
Entrez Gene ID
8881
Gene Name
cell division cycle 16
Gene Aliases
ANAPC6, APC6, CDC16Hs, CUT9
Chromosome Location
Chr.13: 114234845 - 114272675 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078645.1 NP_001072113.1 4 - 5 438 61
NM_001318517.1 NP_001305446.1 4 - 5 435 61
NM_001318518.1 NP_001305447.1 4 - 5 435 61
NM_003903.3 NP_003894.3 4 - 5 438 61
XM_005266206.1 XP_005266263.1 4 - 5 480 61
XM_006719992.1 XP_006720055.1 4 - 5 425 61
XM_006719993.1 XP_006720056.1 3 - 4 373 61
XM_011534851.1 XP_011533153.1 4 - 5 480 61
XM_011534853.1 XP_011533155.1 4 - 5 480 61
XM_017020830.1 XP_016876319.1 4 - 5 480 61
XM_017020831.1 XP_016876320.1 4 - 5 480 61
XM_017020832.1 XP_016876321.1 3 - 4 373 61
XM_017020833.1 XP_016876322.1 2 - 3 58 61
GenBank mRNA AB209850.1 - 3 - 4 1591 61
AF164598.1 - 4 - 5 241 61
AK296424.1 - 4 - 5 428 61
AK315302.1 - 4 - 5 269 61
AL540490.3 - 4 - 5 434 61
BC017244.1 - 4 - 5 348 61
DC323610.1 - 4 - 5 441 61
U18291.1 - 4 - 5 261 61

View Full Product Details