The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005687.4 | NP_005678.3 | 1 - 2 | 341 | 88 |
NR_130154.1 | 1 - 2 | 341 | 88 | ||
XM_006712169.2 | XP_006712232.1 | 1 - 2 | 73 | 88 | |
XM_011510466.2 | XP_011508768.1 | 1 - 2 | 78 | 88 | |
XM_017003110.1 | XP_016858599.1 | 1 - 2 | 75 | 88 | |
GenBank mRNA | AF042346.1 | - | 1 - 2 | 59 | 88 |
AF161521.1 | - | 1 - 2 | 79 | 88 | |
AK001025.1 | - | 1 - 2 | 64 | 88 | |
AK291531.1 | - | 1 - 2 | 76 | 88 | |
AK316497.1 | - | 1 - 2 | 75 | 88 | |
BC006502.2 | - | 1 - 2 | 48 | 88 | |
BC017783.1 | - | 1 - 2 | 71 | 88 | |
BG721840.1 | - | 1 - 2 | 343 | 88 | |
D84430.1 | - | 1 - 2 | 72 | 88 |