Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNPO3
Entrez Gene ID
23534
Gene Name
transportin 3
Gene Aliases
IPO12, LGMD1F, MTR10A, TRN-SR, TRN-SR2, TRNSR
Chromosome Location
Chr.7: 128954180 - 129055173 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012470.3 NP_036602.1 11 - 12 1899 64
NR_034053.2 12 - 13 2060 64
XM_011515989.2 XP_011514291.1 11 - 12 1501 64
GenBank mRNA AF145029.1 - 11 - 12 1711 64
AJ133769.1 - 11 - 12 1496 64
AK001263.1 - 4 - 5 453 64
AK022910.1 - 2 - 3 233 64
AK312580.1 - 11 - 12 1626 64
BC068530.1 - 11 - 12 1627 64

View Full Product Details