Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNPO3
Entrez Gene ID
23534
Gene Name
transportin 3
Gene Aliases
IPO12, LGMD1F, MTR10A, TRN-SR, TRN-SR2, TRNSR
Chromosome Location
Chr.7: 128954180 - 129055173 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012470.3 NP_036602.1 12 - 13 2097 80
NR_034053.2 13 - 14 2258 80
XM_011515989.2 XP_011514291.1 12 - 13 1699 80
GenBank mRNA AF145029.1 - 12 - 13 1909 80
AJ133769.1 - 12 - 13 1694 80
AK001263.1 - 5 - 6 651 80
AK022910.1 - 3 - 4 431 80
AK312580.1 - 12 - 13 1824 80
BC068530.1 - 12 - 13 1825 80

View Full Product Details