Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNPO3
Entrez Gene ID
23534
Gene Name
transportin 3
Gene Aliases
IPO12, LGMD1F, MTR10A, TRN-SR, TRN-SR2, TRNSR
Chromosome Location
Chr.7: 128954180 - 129055173 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191028.2 NP_001177957.2 14 - 15 2134 76
NM_012470.3 NP_036602.1 15 - 16 2326 76
NR_034053.2 16 - 17 2487 76
XM_011515989.2 XP_011514291.1 15 - 16 1928 76
GenBank mRNA AF145029.1 - 15 - 16 2138 76
AJ133769.1 - 15 - 16 1923 76
AK001263.1 - 8 - 9 880 76
AK022910.1 - 6 - 7 660 76
AK225999.1 - 14 - 15 2105 76
AK312580.1 - 15 - 16 2053 76
BC009221.2 - 3 - 4 170 76
BC068530.1 - 15 - 16 2054 76

View Full Product Details