Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNPO3
Entrez Gene ID
23534
Gene Name
transportin 3
Gene Aliases
IPO12, LGMD1F, MTR10A, TRN-SR, TRN-SR2, TRNSR
Chromosome Location
Chr.7: 128954180 - 129055173 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191028.2 NP_001177957.2 1 - 2 521 81
NM_012470.3 NP_036602.1 1 - 2 521 81
GenBank mRNA AF145029.1 - 1 - 2 231 81
AJ133769.1 - 1 - 2 118 81
AK225999.1 - 1 - 2 492 81
AK312580.1 - 1 - 2 248 81
BC068530.1 - 1 - 2 249 81
CB992781.1 - 1 - 2 519 81
DA773316.1 - 1 - 2 503 81

View Full Product Details