Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNPO3
Entrez Gene ID
23534
Gene Name
transportin 3
Gene Aliases
IPO12, LGMD1F, MTR10A, TRN-SR, TRN-SR2, TRNSR
Chromosome Location
Chr.7: 128954180 - 129055173 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191028.2 NP_001177957.2 6 - 7 1272 130
NM_012470.3 NP_036602.1 6 - 7 1272 130
NR_034053.2 7 - 8 1433 130
XM_011515989.2 XP_011514291.1 6 - 7 874 130
GenBank mRNA AF145029.1 - 6 - 7 982 130
AJ133769.1 - 6 - 7 869 130
AK225999.1 - 6 - 7 1243 130
AK312580.1 - 6 - 7 999 130
BC068530.1 - 6 - 7 1000 130

View Full Product Details