Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MASP1
Entrez Gene ID
5648
Gene Name
mannan binding lectin serine peptidase 1
Gene Aliases
3MC1, CRARF, CRARF1, MAP1, MASP, MASP3, MAp44, PRSS5, RaRF
Chromosome Location
Chr.3: 187216085 - 187292220 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031849.2 NP_001027019.1 2 - 3 629 85
NM_001879.5 NP_001870.3 2 - 3 629 85
NM_139125.3 NP_624302.1 2 - 3 629 85
XM_006713701.2 XP_006713764.1 3 - 4 702 85
XM_011512989.2 XP_011511291.1 2 - 3 623 85
XM_011512990.2 XP_011511292.1 2 - 3 623 85
XM_011512991.2 XP_011511293.1 2 - 3 618 85
XM_017006869.1 XP_016862358.1 3 - 4 707 85
XM_017006871.1 XP_016862360.1 2 - 3 623 85
GenBank mRNA AF284421.1 - 2 - 3 329 85
AK129893.1 - 2 - 3 328 85
AK291157.1 - 2 - 3 465 85
AK291686.1 - 2 - 3 307 85
AK300663.1 - 3 - 4 450 85
AL134380.1 - 2 - 3 571 85
BC039724.1 - 2 - 3 564 85
BC106945.1 - 2 - 3 289 85
BC106946.1 - 2 - 3 289 85

View Full Product Details