The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002950.3 | NP_002941.1 | 4 - 5 | 924 | 97 |
GenBank mRNA | AK223595.1 | - | 4 - 5 | 897 | 97 |
AK296905.1 | - | 4 - 5 | 800 | 97 | |
AK297513.1 | - | 4 - 5 | 909 | 97 | |
AK297941.1 | - | 5 - 6 | 793 | 97 | |
AK312369.1 | - | 4 - 5 | 866 | 97 | |
BC007995.1 | - | 4 - 5 | 731 | 97 | |
BC010839.1 | - | 4 - 5 | 885 | 97 | |
CR456742.1 | - | 4 - 5 | 848 | 97 | |
CR749284.1 | - | 4 - 5 | 878 | 97 | |
Y00281.1 | - | 4 - 5 | 985 | 97 |