The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018180.2 | NP_060650.2 | 4 - 5 | 1584 | 70 |
XM_017016404.1 | XP_016871893.1 | 6 - 7 | 1916 | 70 | |
XM_017016405.1 | XP_016871894.1 | 5 - 6 | 1606 | 70 | |
GenBank mRNA | AF427340.1 | - | 4 - 5 | 1585 | 70 |
AK001556.1 | - | 4 - 5 | 1378 | 70 | |
AK001751.1 | - | 4 - 5 | 1557 | 70 | |
AK024869.1 | - | 2 - 3 | 400 | 70 | |
AK315430.1 | - | 4 - 5 | 1199 | 70 | |
AL162051.1 | - | 2 - 3 | 382 | 70 | |
AY064250.1 | - | 4 - 5 | 1552 | 70 | |
BC002473.2 | - | 4 - 5 | 1120 | 70 | |
BC037925.1 | - | 1 - 2 | 904 | 70 | |
BC068471.1 | - | 5 - 6 | 1581 | 70 |