The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018180.2 | NP_060650.2 | 5 - 6 | 1682 | 79 |
XM_017016404.1 | XP_016871893.1 | 7 - 8 | 2014 | 79 | |
XM_017016405.1 | XP_016871894.1 | 6 - 7 | 1704 | 79 | |
GenBank mRNA | AF427340.1 | - | 5 - 6 | 1683 | 79 |
AF427341.1 | - | 2 - 3 | 262 | 79 | |
AK001556.1 | - | 5 - 6 | 1476 | 79 | |
AK001751.1 | - | 5 - 6 | 1655 | 79 | |
AK024869.1 | - | 3 - 4 | 498 | 79 | |
AK315430.1 | - | 5 - 6 | 1297 | 79 | |
AL162051.1 | - | 3 - 4 | 480 | 79 | |
AY064250.1 | - | 5 - 6 | 1650 | 79 | |
BC002473.2 | - | 5 - 6 | 1218 | 79 | |
BC037925.1 | - | 2 - 3 | 1002 | 79 | |
BC068471.1 | - | 6 - 7 | 1679 | 79 |